RIE156321413 Rieke Metals, Inc.

3-Bromo-4'-(thiomethyl)benzophenone

  • MDL number MFCD06201459
  • Cas Number : 197439-23-9
  • Molecular Weight : 307.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5217-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5217-18
Availability details for
Quantity
  • Availability
  • Add to Cart