RIE156321414 Rieke Metals, Inc.

4-Bromo-4'-(thiomethyl)benzophenone

  • MDL number MFCD06201441
  • Cas Number : 197439-28-4
  • Molecular Weight : 307.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5217-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5217-19
Availability details for
Quantity
  • Availability
  • Add to Cart