RIE156321417 Rieke Metals, Inc.

3,5-Difluoro-4'-(thiomethyl)benzophenone

  • MDL number MFCD06201515
  • Cas Number : 197439-19-3
  • Molecular Weight : 264.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5217-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5217-42
Availability details for
Quantity
  • Availability
  • Add to Cart