RIE156321421 Rieke Metals, Inc.

3,3',4,5'-Tetrafluorobenzophenone

  • MDL number MFCD06201522
  • Cas Number : 844885-15-0
  • Molecular Weight : 254.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5192-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5192-42
Availability details for
Quantity
  • Availability
  • Add to Cart