RIE156321422 Rieke Metals, Inc.

4-tert-Butyl-3',4'-(ethylenedioxy)benzophenone

  • MDL number MFCD09801586
  • Molecular Weight : 296.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5202-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5202-34
Availability details for
Quantity
  • Availability
  • Add to Cart