RIE156321423 Rieke Metals, Inc.

5-Fluoro-3,3',4'-trichlorobenzophenone

  • MDL number MFCD06201545
  • Cas Number : 844885-33-2
  • Molecular Weight : 303.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5186-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5186-47
Availability details for
Quantity
  • Availability
  • Add to Cart