RIE156321425 Rieke Metals, Inc.

4-Bromo-3',5'-difluorobenzophenone

  • MDL number MFCD06201446
  • Cas Number : 844879-03-4
  • Molecular Weight : 297.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromophenyl)(3,5-difluorophenyl)methanone
Inchi Code 1S/C13H7BrF2O/c14-10-3-1-8(2-4-10)13(17)9-5-11(15)7-12(16)6-9/h1-7H
Inchi Key NSSPHHIIJGNZTB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5193-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5193-19
Availability details for
Quantity
  • Availability
  • Add to Cart