RIE156321428 Rieke Metals, Inc.

3-Chloro-4'-(ethylthio)benzophenone

  • MDL number MFCD06201493
  • Cas Number : 844884-99-7
  • Molecular Weight : 276.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5203-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5203-15
Availability details for
Quantity
  • Availability
  • Add to Cart