RIE156321429 Rieke Metals, Inc.

4-Chloro-4'-(ethylthio)benzophenone

  • MDL number MFCD06201509
  • Cas Number : 844885-04-7
  • Molecular Weight : 276.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5203-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5203-16
Availability details for
Quantity
  • Availability
  • Add to Cart