RIE156321437 Rieke Metals, Inc.

3-Chloro-3',4'-dimethoxybenzophenone

  • MDL number MFCD06201489
  • Cas Number : 116412-84-1
  • Molecular Weight : 276.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5194-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5194-15
Availability details for
Quantity
  • Availability
  • Add to Cart