RIE156321439 Rieke Metals, Inc.

4-Chloro-3',4'-dimethoxybenzophenone

  • MDL number MFCD02127307
  • Cas Number : 116412-83-0
  • Molecular Weight : 276.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5194-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5194-16
Availability details for
Quantity
  • Availability
  • Add to Cart