RIE156321440 Rieke Metals, Inc.

3,4-Difluoro-4'-(ethylthio)benzophenone

  • MDL number MFCD06201567
  • Cas Number : 845781-10-4
  • Molecular Weight : 278.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5203-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5203-41
Availability details for
Quantity
  • Availability
  • Add to Cart