RIE156321442 Rieke Metals, Inc.

2-chloro-5-picolinoylpyridine

  • MDL number MFCD02260455
  • Cas Number : 884504-81-8
  • Molecular Weight : 218.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5510

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5510
Availability details for
Quantity
  • Availability
  • Add to Cart