RIE156322267 Rieke Metals, Inc.

2-Octanoylpyridine

  • MDL number MFCD07699186
  • Cas Number : 898779-37-8
  • Molecular Weight : 205.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-01F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-01F
Availability details for
Quantity
  • Availability
  • Add to Cart