RIE156322269 Rieke Metals, Inc.

2-Undecanoylpyridine

  • MDL number MFCD07699188
  • Cas Number : 134319-42-9
  • Molecular Weight : 247.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-pyridinyl)-1-undecanone
Inchi Code 1S/C16H25NO/c1-2-3-4-5-6-7-8-9-13-16(18)15-12-10-11-14-17-15/h10-12,14H,2-9,13H2,1H3
Inchi Key NHAZBLPRJWQVIM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5569-01i

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-01i
Availability details for
Quantity
  • Availability
  • Add to Cart