RIE156322270 Rieke Metals, Inc.

2-Dodecanoylpyridine

  • MDL number MFCD07699189
  • Cas Number : 898779-43-6
  • Molecular Weight : 261.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-01J

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-01J
Availability details for
Quantity
  • Availability
  • Add to Cart