RIE156322271 Rieke Metals, Inc.

2-Tridecanoylpyridine

  • MDL number MFCD07699190
  • Cas Number : 242458-31-7
  • Molecular Weight : 275.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-01K

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-01K
Availability details for
Quantity
  • Availability
  • Add to Cart