RIE156322273 Rieke Metals, Inc.

Cyclobutyl 2-pyridyl ketone

  • MDL number MFCD07699201
  • Cas Number : 515154-32-2
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclobutyl(2-pyridinyl)methanone
Inchi Code 1S/C10H11NO/c12-10(8-4-3-5-8)9-6-1-2-7-11-9/h1-2,6-8H,3-5H2
Inchi Key JUFVVUQDXNEDAV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5569-06c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-06c
Availability details for
Quantity
  • Availability
  • Add to Cart