RIE156322277 Rieke Metals, Inc.

1-(2-pyridyl)hexan-1,5-dione

  • MDL number MFCD07699206
  • Cas Number : 246160-12-3
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-07C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-07C
Availability details for
Quantity
  • Availability
  • Add to Cart