RIE156322278 Rieke Metals, Inc.

1-(2-pyridyl)heptan-1,6-dione

  • MDL number MFCD07699207
  • Cas Number : 898779-56-1
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-07D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-07D
Availability details for
Quantity
  • Availability
  • Add to Cart