RIE156322281 Rieke Metals, Inc.

(4-Cyclohexyl)butyl 2-pyridyl ketone

  • MDL number MFCD07699212
  • Cas Number : 898779-64-1
  • Molecular Weight : 245.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-08D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-08D
Availability details for
Quantity
  • Availability
  • Add to Cart