RIE156322283 Rieke Metals, Inc.

1-Methylbutyl 2-pyridyl ketone

  • MDL number MFCD07699216
  • Cas Number : 855377-39-8
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-1-(2-pyridinyl)-1-pentanone
Inchi Code 1S/C11H15NO/c1-3-6-9(2)11(13)10-7-4-5-8-12-10/h4-5,7-9H,3,6H2,1-2H3
Inchi Key TZSWDPXYMIGUQJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5569-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-15
Availability details for
Quantity
  • Availability
  • Add to Cart