RIE156322284 Rieke Metals, Inc.

2-Methylbutyl 2-pyridyl ketone

  • MDL number MFCD07699217
  • Cas Number : 898779-68-5
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-16
Availability details for
Quantity
  • Availability
  • Add to Cart