RIE156322286 Rieke Metals, Inc.

1-Ethylpentyl 2-pyridyl ketone

  • MDL number MFCD07782992
  • Cas Number : 898779-70-9
  • Molecular Weight : 205.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5569-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5569-19
Availability details for
Quantity
  • Availability
  • Add to Cart