RIE156322291 Rieke Metals, Inc.

2-(4-Trifluoromethylbenzoyl)pyridine

  • MDL number MFCD07699230
  • Cas Number : 122377-19-9
  • Molecular Weight : 251.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-10
Availability details for
Quantity
  • Availability
  • Add to Cart