RIE156322292 Rieke Metals, Inc.

2-(3-Bromobenzoyl)pyridine

  • MDL number MFCD07699237
  • Cas Number : 206357-82-6
  • Molecular Weight : 262.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-18
Availability details for
Quantity
  • Availability
  • Add to Cart