RIE156322293 Rieke Metals, Inc.

2-(3-Iodobenzoyl)pyridine

  • MDL number MFCD07699240
  • Cas Number : 898779-80-1
  • Molecular Weight : 309.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-21
Availability details for
Quantity
  • Availability
  • Add to Cart