RIE156322294 Rieke Metals, Inc.

2-(4-Iodobenzoyl)pyridine

  • MDL number MFCD07699241
  • Cas Number : 898779-82-3
  • Molecular Weight : 309.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-22
Availability details for
Quantity
  • Availability
  • Add to Cart