RIE156322296 Rieke Metals, Inc.

2-(4-Cyanobenzoyl)pyridine

  • MDL number MFCD07699243
  • Cas Number : 57954-94-6
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-25

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-25
Availability details for
Quantity
  • Availability
  • Add to Cart