RIE156322297 Rieke Metals, Inc.

2-(3-Phenoxybenzoyl)pyridine

  • MDL number MFCD07699244
  • Cas Number : 898779-86-7
  • Molecular Weight : 275.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-phenoxyphenyl)(2-pyridinyl)methanone
Inchi Code 1S/C18H13NO2/c20-18(17-11-4-5-12-19-17)14-7-6-10-16(13-14)21-15-8-2-1-3-9-15/h1-13H
Inchi Key BDDMYASOHOHAMU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5570-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-27
Availability details for
Quantity
  • Availability
  • Add to Cart