RIE156322302 Rieke Metals, Inc.

2-(4-Butylbenzoyl)pyridine

  • MDL number MFCD07699251
  • Cas Number : 61780-16-3
  • Molecular Weight : 239.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-35C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-35C
Availability details for
Quantity
  • Availability
  • Add to Cart