RIE156322305 Rieke Metals, Inc.

2-(4-Heptylbenzoyl)pyridine

  • MDL number MFCD07699254
  • Cas Number : 898779-98-1
  • Molecular Weight : 281.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-heptylphenyl)(2-pyridinyl)methanone
Inchi Code 1S/C19H23NO/c1-2-3-4-5-6-9-16-11-13-17(14-12-16)19(21)18-10-7-8-15-20-18/h7-8,10-15H,2-6,9H2,1H3
Inchi Key MJQQTRUIJSYSPO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5570-35f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-35f
Availability details for
Quantity
  • Availability
  • Add to Cart