RIE156322309 Rieke Metals, Inc.

2-(4-Hexyloxybenzoyl)pyridine

  • MDL number MFCD07699258
  • Cas Number : 898780-09-1
  • Molecular Weight : 283.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-36E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-36E
Availability details for
Quantity
  • Availability
  • Add to Cart