RIE156322310 Rieke Metals, Inc.

2-(4-Heptyloxybenzoyl)pyridine

  • MDL number MFCD07699259
  • Cas Number : 898780-12-6
  • Molecular Weight : 297.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-36F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-36F
Availability details for
Quantity
  • Availability
  • Add to Cart