RIE156322312 Rieke Metals, Inc.

2-(2,4-Difluorobenzoyl)pyridine

  • MDL number MFCD07699261
  • Cas Number : 898780-18-2
  • Molecular Weight : 219.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-difluorophenyl)(2-pyridinyl)methanone
Inchi Code 1S/C12H7F2NO/c13-8-4-5-9(10(14)7-8)12(16)11-3-1-2-6-15-11/h1-7H
Inchi Key ZVYATEOQCVIQDD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5570-38

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-38
Availability details for
Quantity
  • Availability
  • Add to Cart