RIE156322313 Rieke Metals, Inc.

2-(2,5-Difluorobenzoyl)pyridine

  • MDL number MFCD07699262
  • Cas Number : 898780-21-7
  • Molecular Weight : 219.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-39

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-39
Availability details for
Quantity
  • Availability
  • Add to Cart