RIE156322314 Rieke Metals, Inc.

2-(2,6-Difluorobenzoyl)pyridine

  • MDL number MFCD07699263
  • Cas Number : 898780-24-0
  • Molecular Weight : 219.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-40

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-40
Availability details for
Quantity
  • Availability
  • Add to Cart