RIE156322326 Rieke Metals, Inc.

2-(3,4-Dimethylbenzoyl)pyridine

  • MDL number MFCD07699279
  • Cas Number : 102001-19-4
  • Molecular Weight : 211.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-59
Availability details for
Quantity
  • Availability
  • Add to Cart