RIE156322328 Rieke Metals, Inc.

2-(3,4,5-Trifluorobenzoyl)pyridine

  • MDL number MFCD07699281
  • Cas Number : 898758-18-4
  • Molecular Weight : 237.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5570-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-61
Availability details for
Quantity
  • Availability
  • Add to Cart