RIE156322329 Rieke Metals, Inc.

2-Butyryloxazole

  • MDL number MFCD07699284
  • Cas Number : 898758-24-2
  • Molecular Weight : 139.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-01B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-01B
Availability details for
Quantity
  • Availability
  • Add to Cart