RIE156322332 Rieke Metals, Inc.

Hexyl oxazol-2-yl ketone

  • MDL number MFCD07699287
  • Cas Number : 898758-33-3
  • Molecular Weight : 181.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-01E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-01E
Availability details for
Quantity
  • Availability
  • Add to Cart