RIE156322333 Rieke Metals, Inc.

2-Octanoyloxazole

  • MDL number MFCD07699288
  • Cas Number : 898758-36-6
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-01F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-01F
Availability details for
Quantity
  • Availability
  • Add to Cart