RIE156322336 Rieke Metals, Inc.

2-Undecanoyloxazole

  • MDL number MFCD07699291
  • Cas Number : 898758-45-7
  • Molecular Weight : 237.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(1,3-oxazol-2-yl)-1-undecanone
Inchi Code 1S/C14H23NO2/c1-2-3-4-5-6-7-8-9-10-13(16)14-15-11-12-17-14/h11-12H,2-10H2,1H3
Inchi Key ZXRINHMDCFEPIR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5571-01i

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-01i
Availability details for
Quantity
  • Availability
  • Add to Cart