RIE156322340 Rieke Metals, Inc.

2-(5-Chlorovaleryl)oxazole

  • MDL number MFCD07699307
  • Cas Number : 898758-80-0
  • Molecular Weight : 187.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-05D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-05D
Availability details for
Quantity
  • Availability
  • Add to Cart