RIE156322342 Rieke Metals, Inc.

2-Cyclopentanoyloxazole

  • MDL number MFCD07699310
  • Cas Number : 898758-86-6
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-06D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-06D
Availability details for
Quantity
  • Availability
  • Add to Cart