RIE156322346 Rieke Metals, Inc.

2-(6-Oxoheptanoyl)oxazole

  • MDL number MFCD07699317
  • Cas Number : 898759-00-7
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-07D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-07D
Availability details for
Quantity
  • Availability
  • Add to Cart