RIE156322347 Rieke Metals, Inc.

2-(Cyclohexylacetyl)oxazole

  • MDL number MFCD07699319
  • Cas Number : 898759-04-1
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-08A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-08A
Availability details for
Quantity
  • Availability
  • Add to Cart