RIE156322348 Rieke Metals, Inc.

2-(3-Cyclohexylpropionyl)oxazole

  • MDL number MFCD07699320
  • Cas Number : 898759-06-3
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-cyclohexyl-1-(1,3-oxazol-2-yl)-1-propanone
Inchi Code 1S/C12H17NO2/c14-11(12-13-8-9-15-12)7-6-10-4-2-1-3-5-10/h8-10H,1-7H2
Inchi Key WMWFDVDGCXETNM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5571-08b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-08b
Availability details for
Quantity
  • Availability
  • Add to Cart