RIE156322350 Rieke Metals, Inc.

2-(5-Cyclohexylvaleryl)oxazole

  • MDL number MFCD07699322
  • Cas Number : 898759-10-9
  • Molecular Weight : 235.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-08D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-08D
Availability details for
Quantity
  • Availability
  • Add to Cart