RIE156322352 Rieke Metals, Inc.

2-(2,2,2-Trimethylacetyl)oxazole

  • MDL number MFCD07699324
  • Cas Number : 898759-14-3
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5571-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5571-11
Availability details for
Quantity
  • Availability
  • Add to Cart